GSK-3
GSK-3 is a highly conserved family of protein kinases. In humans, two genes encode two distinct, but closely related GSK-3 forms, referred to as GSK-3α and GSK-3β. They display 84% overall identity and 98% identity within their catalytic domain, the main difference between the two coming from an extra Gly-rich stretch in the N terminal domain of GSK-3α. Yet they are not functionally interchangeable as demonstrated by the embryonic lethal phenotype of GSK-3β knock-outs. GSK-3β2 is an alternate splicing variant of GSK-3ßβ expressed in brain.
GSK-3β has been crystallized with various ligands. Its overall shape is shared by all kinases, and comprises a small, N-terminal lobe consisting mostly of β sheets and a large, C-terminal lobe, essentially formed of α-helices. The ATP-binding pocket is located between the two lobes. Arg96, Arg180 and Lys205 form a small pocket where the phosphate group of the primed substrate and the pseudo-substrate binds.
GSK-3α/β is regulated by phosphorylation on Ser21/9 (inhibitory) and on Tyr279/216 (activating). The phospho-Ser21/9 N-terminal domain of GSK-3 acts as a pseudo-substrate blocking access of substrates to the catalytic site. The T-loop domain containing unphosphorylated Tyr279/216 is thought to prevent access of substrates to the catalytic site, and Tyr phosphorylation to release this inhibition. GSK-3 is also regulated by interactions with other proteins. Axin and presenilin act as docking proteins that allow the substrate to interact with the priming kinase. The substrate indeed requires a priming phosphorylation by another kinase on a residue located C-terminal to the GSK-3 phosphorylation site. The substrate recognition site of GSK-3 is -S-X-X-X-Sp-, where Sp is the priming pre-phosphorylated Ser. GSK-3β is a predominantly cytoplasmic kinase, but it is also present in the nucleus and in mitochondria. FRAT-1 promotes the nuclear efflux of GSK-3β. LANA, a Kaposi virus protein, sequesters GSK-3β in the nucleus.
GSK-3 plays a major function in Wnt signalling pathways. When the Wnt pathway is stimulated, GSK-3 is inactivated, β-catenin builds up and accumulates in the nucleus where it forms with TCF/LEF, a transcription factor regulating a large variety of genes. GSK-3 phosphorylates the cell cycle regulators β-catenin, cyclin D1, cyclin E, p21CIP1 and c-Myc, leading to their ubiquitin-dependent destruction. In the absence of insulin, active GSK-3 phosphorylates and inactivates glycogen synthase and eIF2B. Binding of insulin to its plasma membrane receptor leads to PKB/AKT activation, resulting in phosphorylation and inactivation of GSK-3. Consequently, glycogen synthase and eIF2B are activated, and glycogen and protein synthesis are stimulated. GSK-3 plays a pro-apoptotic function in neuronal cells. In contrast, GSK-3 is necessary also for cell survival as demonstrated by the massive TNFα-induced hepatocyte apoptosis which leads to death in early embryonic GSK-3β knockout mice. GSK-3 activity/inactivity or spatial distribution play an essential role during development (polarity determination). Finally, GSK-3 and CK1 regulate the circadian clock in Drosophila and in mammals.
Five sets of data have stimulated altogether the search for pharmacological inhibitors of GSK-3: (i) the mood stabilizing properties of lithium, the first GSK-3 inhibitor to be described; (ii) the insulin-mimetic properties of GSK-3 inhibitors; (iii) the interaction of GSK-3 with presenilin-1, the GSK-3-dependent amyloid-β production and abnormal tau phosphorylation in Alzheimer's disease; (iv) the involvement of GSK-3 in neuronal cell death and the neuroprotection provided by GSK-3 inhibitors following various insults; and (v) the maintenance of pluripotency of embryonic stem cells in the absence of feeder cells by GSK-3 inhibitors. Over 30 GSK-3 inhibitors have been identified, among which seven have been co-crystallized with GSK-3β, all of which localize within the ATP-binding pocket of the enzyme. GSK-3 inhibitors are thus evaluated on Alzheimer's disease and other neurodegenerative diseases, bipolar affective disorders, diabetes and diseases caused by unicellular parasites.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Family Members GSK-3α GSK-3β GSK-3β2
Other Names Glycogen synthase kinase 3α Glycogen synthase kinase 3β Glycogen synthase kinase 3β2
Molecular Weight (kDa) 51 kDa 47 kDa 48 kDa
Structural Data 483 aa (human) 420 aa (human) 433 aa (human)
Isoforms Not Known Not Known Splice variant of GSK-3β
Species Present in all species Present in all species   Not Known
Domain Organization Not Known Not Known Not Known
Phosphorylation Sites Ser2
Tyr279
Ser9
Tyr216
Not Known
Tissue Distribution Ubiquitous Ubiquitous Brain
Subcellular Localization Cytoplasm, nucleus, mitochondria Cytoplasm, nucleus, mitochondria Cytoplasm, nucleus, mitochondria
Binding Partners/ Associated Proteins Axin
Presenilin
β-catenin
Akt
p53
APC
Tau
Notch2
Glycogen synthase
Axin
Presenilin
β-catenin
Akt
p53
APC
Tau
Notch2
Glycogen synthase
Axin
Presenilin
β-catenin
Akt
p53
APC
Tau
Notch2
Glycogen synthase
Upstream Activators CK2 (for glycogen synthase)
CK1α (β-catenin)
DYRK1A (eIF2B)
PKA (Cubitus interruptus)
CDK5/p25 (tau)
CK2 (for glycogen synthase)
CK1α (β-catenin)
DYRK1A (eIF2B)
PKA (Cubitus interruptus)
CDK5/p25 (tau)
CK2 (for glycogen synthase)
CK1α (β-catenin)
DYRK1A (eIF2B)
PKA (Cubitus interruptus)
CDK5/p25 (tau)
Downstream Activation p70S6K
p90rsk
PKA
Akt
PKC isoforms
ILK
fyn kinase
PYK2
ZAK1
MEK1
GSK-3
MAP-IB
MAP-2
eIF2B
Cubitus interruptus
p70S6K
p90rsk
PKA
Akt
PKC isoforms
ILK
fyn kinase
PYK2
ZAK1
MEK1
GSK-3
MAP-IB
MAP-2
eIF2B
Cubitus interruptus
p70S6K
p90rsk
PKA
Akt
PKC isoforms
ILK
fyn kinase
PYK2
ZAK1
MEK1
GSK-3
MAP-IB
MAP-2
eIF2B
Cubitus interruptus
Activators Not Known Not Known Not Known
Inhibitors SB 216763 (S3442)
SB 415286 (S3567)
FRAT1/FRAT2 (GBP)
Lithium (213233)
Kenpaullone (K3888)
Alsterpaullone (A4847)
Indirubin-3’-oxime (I0404)
6-bromo-indirubin-3’-oxime (BIO) (B1686)
Hymenialdisine
Aloisine
TDZD-8 (T8325)
Ro 31-8220 (bisindolylmaleimide IX) (R136)
FRATtide (FRAT1188-226 peptide)
GID320-429 and GID380-404 peptides
SB 216763 (S3442)
SB 415286 (S3567)
FRAT1/FRAT2 (GBP)
Lithium (213233)
Kenpaullone (K3888)
Alsterpaullone (A4847)
Indirubin-3’-oxime (I0404)
6-bromo-indirubin-3’-oxime (BIO) (B1686)
Hymenialdisine
Aloisine
TDZD-8 (T8325)
Ro 31-8220 (bisindolylmaleimide IX) (R136)
FRATtide (FRAT1188-226 peptide)
GID320-429 and GID380-404 peptides
SB 216763 (S3442)
SB 415286 (S3567)
FRAT1/FRAT2 (GBP)
Lithium (213233)
Kenpaullone (K3888)
Alsterpaullone (A4847)
Indirubin-3’-oxime (I0404)
6-bromo-indirubin-3’-oxime (BIO) (B1686)
Hymenialdisine
Aloisine
TDZD-8 (T8325)
Ro 31-8220 (bisindolylmaleimide IX) (R136)
FRATtide (FRAT1188-226 peptide)
GID320-429 and GID380-404 peptides
Selective Activators Not Known Not Known Not Known
Physiological Function Wnt and Hedgehog signaling pathways, transcription, cell division cycle regulation response to DNA damage, cell death and cell survival, cardiovascular and neuronal functions, insulin transduction development, differentiation, regulation of circadian rhythm, stem cell pluripotency maintenance and differentiation Wnt and Hedgehog signaling pathways, transcription, cell division cycle regulation response to DNA damage, cell death and cell survival, cardiovascular and neuronal functions, insulin transduction development, differentiation, regulation of circadian rhythm, stem cell pluripotency maintenance and differentiation Wnt and Hedgehog signaling pathways, transcription, cell division cycle regulation response to DNA damage, cell death and cell survival, cardiovascular and neuronal functions, insulin transduction development, differentiation, regulation of circadian rhythm, stem cell pluripotency maintenance and differentiation
Disease Relevance Bipolar disorder, Alzheimer's disease, stroke, Parkinson's disease, Huntington's disease, Transmissible spongiform encephalopathies, schizophrenia, circadian clock-dependent diseases, Type 2 diabetes, prostate cancer Bipolar disorder, Alzheimer's disease, stroke, Parkinson's disease, Huntington's disease, Transmissible spongiform encephalopathies, schizophrenia, circadian clock-dependent diseases, Type 2 diabetes, prostate cancer Bipolar disorder, Alzheimer's disease, stroke, Parkinson's disease, Huntington's disease, Transmissible spongiform encephalopathies, schizophrenia, circadian clock-dependent diseases, Type 2 diabetes, prostate cancer
Abbreviations
SB 216763: 3-(2,4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione
SB 415286: 3-[(3-Chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-1H-pyrrol-2,5-dione
Similar Products
Refertences