Potential Application for Anisyl Butyrate
Available in Natural (US) & Kosher
Anisyl butyrate can be used sparingly as an ingredient in condensed milk and cream flavors. Its unique sweetness would find good application in confection, oral care products and as a bitterness modifier.
Key Organoleptic Characteristics   Sweet, anisic with fruity confectionary nuances
Potential Applications Anise spice blends; candy flavors such as red licorice, coconut, vanilla and cream; minor nuance effects in various fruit flavors and oral care products
Natural Occurrence Anise seed oil
Aroma Characteristics @1.0% in ethanol. Sweet anisic, fatty creamy with a candy-like sweetness
Taste Characteristics @5-10 ppm. Sweet creamy, candy-like with good mouth feel
Product Number W210000