Ce-MDIP1 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Ce-MDIP1. 
Applications
  • Catalysis:  Heterogeneous catalysis
Properties
  • Chiral Channels = 10.5 Å x 6.0 Å