CPM-24 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare CPM-24. 
Applications
  • Gas adsorption:  Carbon dioxide, nitrogen gas
Properties
  • Maximum Surface Area: 296 m2 / g Langmuir Surface Area
  • Pore volume = 0.100 cm3 / g