HZIF-1Mo Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare HZIF-1Mo. 
Applications
  • Catalysis:  Oxidation of benzyl alcohol
  • Gas adsorption
Properties
  • Maximum Surface Area: 476 m2 / g Langmuir Surface Area