Mg3(BPT)2(H2O)4 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Mg3(BPT)2(H2O)4
Applications
  • Gas adsorption:  Hydrogen gas
  • Luminescence
Properties
  • Maximum Surface Area: 837 m2 / g Langmuir Surface Area
  • Channels = 11.2 Å × 15.0 Å