MIL-53(Fe) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MIL-53(Fe). 
Applications
When constructed with:
Linker: TPA, H2BDC, BDC
  • Liquid Adsorption: Methanol, ethanol, propanol
Linker: H4DOBDC
  • Separations: Organic substances
  • Gas Adsorption
Properties
When constructed with:
Linker: TPA, H2BDC, BDC
  • none
Linker: H4DOBDC
  • Maximum Surface Area: 1500 m2 / g Langmuir Surface Area