MOF-14 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-14. 
Applications
  • Chemical Synthesis:  Biological assembly
Properties
  • Maximum Surface Area: 1502 m2 / g BET surface area
  • Pore volume = 0.53 cm3 / g
Materials
References
  1. Chen, B.; Eddaoudi, M.; Hyde, S.; O'Keeffe M.; Yaghi, M. Science 2001, 291(5506), 1021-1023.
Related Links