MOF-205 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-205. 
Applications
  • Gas adsorption:  Methane, carbon dioxide gas
  • Separations
Properties
  • Maximum Surface Area: 6170 m2 / g Langmuir Surface Area
  • Pore volume = 0.38 cm3 / g