MOF-205 (DUT-6) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare MOF-205 (DUT-6). 
Applications
  • Gas adsorption:  Hydrogen, nitrogen, methane gas
Properties
  • Maximum Surface Area: 4460 m2 / g BET surface area
Materials
References
  1. Furukawa, H.; Ko, N.; Go, Y.; Aratani, N.; Choi, S.; Choi, E.; Yazaydin, A.; Snurr, R.; O'Keeffe, M.; Kim, J. Science 2010, 329(5990), 424-428.
Related Links