PCN-131' Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare PCN-131'. 
Applications
  • Gas adsorption
Properties
  • Maximum Surface Area: 442 m2 / g BET surface area