SNU-M11 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare SNU-M11. 
Applications
  • Gas adsorption:  Carbon dioxide gas
Properties
  • Maximum Surface Area: 505 m2 / g BET surface area, carbon dioxide gas