UTSA-38 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare UTSA-38. 
Applications
  • Gas adsorption:  Hydrogen gas
  • Catalysis
Properties
  • Maximum Surface Area: 1690 m2 / g Langmuir Surface Area
  • Pore volume = 0.61 cm3 / g