YO - MOF Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare YO - MOF. 
Applications
  • Gas adsorption:  Carbon dioxide, nitrogen, argon gas
Properties
  • Maximum Surface Area: 690 m2 / g Langmuir Surface Area
Materials
References
  1. Mulfort, K. L.; Farha, O. K.; Malliakas, C. D.; Kanatzidis, M. G.; Hupp, J. T.; Chem. Eur. J. 2010, 16(1), 276.
Related Links