ZIF-70 Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare ZIF-70. 
Applications
  • Separations: Carbon monoxide, carbon dioxide gas
Properties
  • Maximum Surface Area: 1970 m2 / g Langmuir Surface Area
  • Pore diameter = 15.9 Å
Materials
References
  1. Banerjee, D.; Phan, B.; Knobler, C.; Furukawa, H.; O'Keeffe, M.; Yaghi, O. Science 2008, 319(5865), 939-943.
  2. Banerjee, R.; Furukawa, H.; Britt, D.; Knobler, C.; O'Keeffe, M.; Yaghi, O. J. Am. Chem. Soc. 2009, 131(11), 3875-3877.
Related Links