Zn3(BTC)2·12H2O Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Zn3(BTC)2·12H2O. 
Applications
  • Liquid adsorption:  Water, ammonia
Properties
  • Density = 1.946 g / cm3
Materials
References
  1. Yaghi, O.; Li, H.; Groy, T. J. Am. Chem. Soc. 1996, 118, 9096-9101.
Related Links