Zn(Im)(aIm) Supplemental Information
Explore MOF Applications, Properties and Materials to construct MOFs in the MOF Constructor Tool
Literature suggests that the following materials can be used to prepare Zn(Im)(aIm). 
Applications
  • Gas adsorption
Properties
  • Maximum Surface Area: 580 m2 / g BET surface area
  • Pore volume = 0.26 cm3 / g
Materials
References
  1. Bernt, S.; Feyand, M.; Modrow, A.; Wack, J.; Senker, J.; Stock, N. Eur. J. Inorg. Chem. 2011, 5378-5383.
Related Links