Fast GC Analysis of Detailed cis/trans Fatty Acid Methyl Esters (FAMEs) on the 75 m SP™-2560 Capillary Column
By: Michael D. Buchanan, Reporter US Vol 25.4
Michael D. Buchanan
mike.buchanan@sial.com
Trans Fat Analysis
Because of the adverse health effects of trans fats, the United States Food and Drug Administration (FDA) requires that food manufacturers list trans fat content on the food’s Nutrition Facts panel (1,2). These labeling requirements have placed added pressure on food analysts to process more samples, which in turn creates the need for a rapid analytical method.
back to top
SP-2560: The Best GC Phase Available for Detailed cis/trans FAME Analyses
Cis/trans selectivity increases with increasing column polarity (percentage of biscyanopropyl). The 100 m x 0.25 mm internal diameter (I.D.), 0.20 μm SP-2560 column is the longest, most polar column currently available. By combining both selectivity of the phase and column efficiency (by virtue of long column length), highly polar 100% biscyanopropyl SP-2560 capillary GC columns provide high resolution cis/trans FAME isomer separation. The SP-2560 column is specified in the Association of Official Analytical Chemists (AOAC) cis/trans FAME method (3).
back to top
Analytical Challenge: Improved Throughput of Detailed cis/trans FAME Analyses
To increase throughput of the detailed cis/trans FAME analysis, Fast GC principles were applied by reducing column length, column I.D., film thickness, and carrier gas viscosity. The result is a significant reduction in analysis time compared to the 100 m column method: 30% reduction of the 37-component FAME sample (Figures 1 and 2), and nearly 50% reduction of the detailed analysis of the C18 isomer mix (Figures 3 and 4). Note that in both cases, peak shape and resolution does not suffer, even with the shorter analysis times.
Figure 1. 37-Component FAME Mix on the 100 m SP-2560 Column (24056)
Figure 2. 37-Component FAME Mix on the 75 m SP-2560 Column (23348-U)
Figure 3. Detailed Analysis of C18 FAME Isomers on the 100 m SP-2560 Column (24056)
Figure 4. Detailed Analysis of C18 FAME Isomers on the 75 m SP-2560 Column (23348-U)
Peak IDs for Figures 1 and 2
In both examples shown here, the loss of total theoretical plates by reducing the column length from 100 m to 75 m is offset by the narrower column I.D. (0.18 vs. 0.25 mm), thinner film (0.14 vs. 0.20 μm) and the higher diffusivity, lower viscosity carrier gas (hydrogen vs. helium). Simply put, the 75 m x 0.18 mm I.D. SP-2560 column does what the 100 m x 0.25 mm I.D. column does, but in a much shorter time. The 0.18 mm I.D. column is compatible with both conventional and Fast GC instruments.
back to top
Conclusion
Supelco offers food analysts performing detailed cis/ trans FAME analyses solutions in terms of both resolving power and speed. The 100 m SP-2560 column provides excellent resolution and is a workhorse column for detailed cis/trans FAME analyses. Now, for analysts interested in improving throughput, a Fast GC version SP-2560 column in 75 m x 0.18 mm, 0.14 μm dimensions, offered exclusively by Supelco, provides both the high resolution and high speed needed to achieve high throughput with detailed cis/trans FAME analyses.
back to top
Materials
References
  1. Ascherio, A.; Willett, W. C. Health effects of trans fatty acids. Am. J. Clin. Nutr. 1997, 66 (suppl.), 1006S-1010S.
  2. US Food and Drug Administration. Questions and Answers about trans Fat Nutrition Labeling. http://www.cfsan.fda.gov/~dms/qatrans2.html. Accessed May 17, 2007.
  3. Official Methods of Analysis of AOAC International, 17th edition, Revision 1 (2002).