Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Acid Fast Stain
Reagent Preparation
Malachite Green solution contains both counterstain and differentiator and is performed within the same step.
Procedure
  1. Deparaffinize tissue sections and hydrate to deionized water.
  2. Stain in Carbol-Fuchsin solution for 30-45 minutes.
  3. Wash in deionzed water to remove excess stain.
  4. Differentiate and counterstain in Malachite Green solution for 2 minutes.
  5. Rinse in tap water and let air dry.
  6. Do not dehydrate slides through graded alcohols after staining.
  7. Dip slides individually in xylene and coverslip.
Results
Acid fast bacteria – Red
Background – Green to turquoise
Acid Fast Stain
Reagents
Related Links