Life Science
Lentiviral Magnetic Transduction
In this exercise, the hard to transduce human melanoma cell line, CRL-1676 will be transduced with MISSION™ TurboGFP Control Transduction particles coupled with ExpressMag magnetic beads as a solution to delivering DNA to hard-to-transduce cells. CRL-1676 cells will also be transduced with a standard polybrene approach that will allow for the comparison of transduction efficiency using both methods.