|
Printer friendly PDF version (416 Kb)
|
 |
| Sigma-Aldrich® HEPES Physical Properties |
| Synonyms |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) |
 |
| Molecular Formula |
C8H18N2O4S |
Beilstein Registry |
883043 |
pKa (at 20) |
7.55 |
| Molecular Weight |
238.30 |
MDL Number |
MFCD00006158 |
pKa (at 25) |
7.48 |
| CAS Number |
7365-45-9 |
EG/EC Number |
230-907-9 |
pKa (at 37) |
7.31 |
| Useful pH range |
6.8 - 8.2 |
|
| Sigma-Aldrich HEPES Specifications Comparison |
| |
H3375 |
H6147 |
H4034 |
H7523 |
54459 |
54457 |
| Grade |
≥99.5% (titration) |
BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration) |
BPC, ≥99.5% (titration), cell culture tested |
BioXtra, pH 5.0 - 6.5 (20 °C, 1 M in H2O), ≥99.5% (titration) |
BioUltra, ≥99.5% (T) |
BioUltra, for molecular biology, ≥99.5% (T) |
| Purity |
≥99.5% |
≥99.5% |
≥99.5% |
≥99.5% |
≥99.5% |
≥99.5% |
| Reagent Testing |
| |
H3375 |
H6147 |
H4034 |
H7523 |
54459 |
54457 |
Enhanced Quality Profile Buffers |
X |
X |
X |
X |
|
|
| 260NM |
|
|
≤0.05 |
≤0.045 |
≤0.045 |
≤0.045 |
| 280NM |
|
|
≤0.05 |
≤0.035 |
≤0.035 |
≤0.035 |
| 290NM |
≤0.05 |
≤0.05 |
≤0.05 |
|
|
|
| IR |
X |
|
X |
|
|
|
| pH |
|
|
|
5.0 - 6.5 (20 °C, 1 M in H2O) |
5.0 - 6.5 (25 °C, 1 M in H2O) |
5.0 - 6.5 (25 °C, 1 M in H2O) |
| Molarity |
|
| Osmolality |
|
| Karl Fischer |
≤0.1% |
≤0.1% |
≤0.1% |
≤0.1% |
|
|
| LOD |
|
|
|
≤1.0% |
≤0.5% |
≤0.5% |
| Insoluble Matter |
|
|
|
filter test |
filter test |
filter test |
| Residue on Ignition |
|
|
|
≤0.1% |
≤0.1% |
≤0.1% |
| Life Science Testing |
| |
H3375 |
H6147 |
H4034 |
H7523 |
54459 |
54457 |
| Animal Component Free |
|
| Endotoxin |
|
|
≤0.1 eu/mg |
|
|
|
| SPC |
|
|
≤100 cfu/g |
|
|
|
| Cell test |
|
|
X |
|
|
|
| Sterility |
|
| RNAse |
|
|
X |
|
|
X |
| DNAse |
|
|
X |
|
|
X |
| Nickase |
|
|
X |
|
|
|
| Protease |
|
|
X |
|
|
X |
| Phosphatase |
|
X |
| Embryo |
|
X |
|
| Trace Elemental Impurities |
| |
H3375 |
H6147 |
H4034 |
H7523 |
54459 |
54457 |
| Sodium(Na) |
|
|
|
≤0.005% |
≤0.0050% |
≤0.0050% |
| Chloride(Cl) |
|
|
|
≤0.05% |
≤0.0050% |
≤0.0050% |
| Sulfate(SO4) |
|
≤0.0200% |
≤0.0050% |
| Aluminum(Al) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Arsenic(As) |
|
|
|
≤0.0001% |
≤0.00001% |
≤0.00001% |
| Barium(Ba) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Bismuth(Bi) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Calcium(Ca) |
|
|
|
≤0.001% |
≤0.0010% |
≤0.0010% |
| Cadmium(Cd) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Cobalt(Co) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Chromium(Cr) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Copper(Cu) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Iron(Fe) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Potassium(K) |
|
|
|
≤0.005% |
≤0.0050% |
≤0.0050% |
| Lithium(Li) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Magnesium(Mg) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Manganese(Mn) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Molybdenum(Mo) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Nickel(Ni) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Strontium(Sr) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Zinc(Zn) |
|
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| |
back to top back to Biological Buffers |