|
Biological Buffers
Trizma® HCl
|
|
|
| Sigma-Aldrich® Trizma® HCl Physical Properties |
| Synonyms |
TRIS HCl TRIS hydrochloride Tris(hydroxymethyl)aminomethane hydrochloride |
 |
| Molecular Formula |
NH2C(CH2OH)3 · HCl |
Beilstein Registry |
3675235 |
Useful pH Range |
7.0 - 9.0 |
| Molecular Weight |
157.60 |
MDL Number |
MFCD00012590 |
pKa (at 25) |
8.1 |
| CAS Number |
1185-53-1 |
EG/EC Number |
214-684-5 |
|
|
| Sigma-Aldrich Trizma HCl Specifications Comparison |
| |
T3253 |
T5941 |
T6666 |
93287 |
93363 |
| Grade |
reagent grade, ≥99.0% (redox titration), crytalline |
for molecular biology, BPC, cell culture tested, ≥99.0% (titration) |
BioXtra, pH 3.5 - 5.0 (20 °C, 0.5 M in H2O), ≥99.0% (titration) |
BioUltra, for luminescence, ≥99.0% (AT) |
BioUltra, for molecular biology, ≥99.0% (AT) |
| Purity |
≥99.0% |
≥99.0% |
≥99.0% |
≥99.0% |
≥99.0% |
| Reagent Testing |
| |
T3253 |
T5941 |
T6666 |
93287 |
93363 |
Enhanced Quality Profile Buffers |
X |
X |
X |
|
|
| λex |
|
|
|
200 - 600 nm |
|
| λem |
|
|
|
300 - 800 nm |
|
| 260NM |
|
≤0.06 |
≤0.015 |
≤0.12 |
≤0.12 |
| 280NM |
|
≤0.06 |
≤0.010 |
≤0.08 |
≤0.08 |
| 290NM |
≤0.05 |
≤0.05 |
|
|
|
| IR |
X |
X |
X |
|
|
| pH |
|
|
3.5 - 5.0 (25 °C, 0.5 M in H2O) |
2.5 - 4.0 (25 °C, 4 M in H2O) |
2.5 - 4.0 (25 °C, 4 M in H2O) |
| Karl Fischer |
≤0.5% |
≤0.5% |
|
|
|
| LOD |
|
|
≤0.2% |
≤0.2% |
≤0.2% |
| Insoluble Matter |
|
|
filter test |
filter test |
filter test |
| Residue on Ignition |
|
|
≤0.2% |
≤0.2% |
≤0.2% |
| Life Science Testing |
| |
T3253 |
T5941 |
T6666 |
93287 |
93363 |
| Endotoxin |
|
≤0.1 eu/mg |
|
|
|
| SPC |
|
≤100 cfu/g |
|
|
|
| Cell test |
|
X |
|
|
|
| RNAse |
|
X |
|
|
X |
| DNAse |
|
X |
|
|
X |
| Nickase |
|
X |
|
|
|
| Protease |
|
X |
|
|
X |
| Phosphatase |
|
X |
| Suitability Electrophoresis |
|
X |
|
|
|
| Trace Elemental Impurities |
| |
T3253 |
T5941 |
T6666 |
93287 |
93363 |
| Sodium(Na) |
|
|
≤0.005% |
≤0.0050% |
≤0.0050% |
| Sulfate(SO4) |
|
|
≤0.005% |
≤0.0050% |
≤0.0050% |
| Aluminum(Al) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Arsenic(As) |
|
|
≤0.0001% |
≤0.00001% |
≤0.00001% |
| Barium(Ba) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Bismuth(Bi) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Calcium(Ca) |
|
|
≤0.001% |
≤0.0010% |
≤0.0010% |
| Cadmium(Cd) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Cobalt(Co) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Chromium(Cr) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Copper(Cu) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Iron(Fe) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Potassium(K) |
|
|
≤0.005% |
≤0.0050% |
≤0.0050% |
| Lithium(Li) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Magnesium(Mg) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Manganese(Mn) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Molybdenum(Mo) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Nickel(Ni) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Strontium(Sr) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| Zinc(Zn) |
|
|
≤0.0005% |
≤0.0005% |
≤0.0005% |
| |
back to top back to Biological Buffers |