Life Science > Zinc Finger Nuclease (ZFN)
Functional Genomics & RNAi
Zinc Finger Nuclease (ZFN)
CompoZr® Zinc Finger Nuclease Technology
Zinc finger nucleases (ZFNs) are a class of engineered DNA-binding proteins that facilitate targeted editing of the genome by creating double-strand breaks in DNA at user-specified locations.
Double-strand breaks are important for site-specific mutagenesis in that they stimulate the cell’s natural DNA-repair processes, namely homologous recombination and Non-Homologous End Joining (NHEJ).
Using well-established and robust protocols, these cellular processes can be harnessed to generate precisely targeted genomic edits resulting in cell lines, including somatic cell lines, with targeted gene deletions (Knockouts), integrations, or modifications.
Method of the Year 2011 – Nature Methods
“Genome Editing with Engineered Nucleases”
For its annual choice to highlight an important research method for biological researchers Nature Methods has selected “Genome Editing with Engineered Nucleases” - Nat Methods. 2011 Dec 28; 9(1):26. In 2011 there were many high profile publications using zinc finger nucleases with a wide range of genomic edits in a variety of cell lines and species, see Zinc Finger Nuclease References.
2011 was also an important year as the leading genome editing technology, CompoZr ZFNs, had a reduction in price that the increased access of the most well-characterized and highly published genome editing technology in the market. Along with the reduction in price Knockout ZFNs are now available on the Sigma website for all human, mouse and rat genes providing easy access to the most common genome editing research application.
CompoZr ZFN Products
Knockout ZFNs   Custom ZFN Service   Targeted Integration Kits
Learning Center:
Tool Box:
Applications Center:
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
 YFG Information
 Your Favorite Gene Search