Head HTML-
Endoplasmic Reticulum
Submitted by: Mr Fabrice Caudron
King's College London
Live cells were stained with DiOC6, washed twice with culture medium (synthetic complete lacking uracile) and placed on an agar pad containing growth medium and DiOC6. A stack was acquired and images were further deconvolved. This image shows a single focal plane of the yeast endoplasmic reticulum stained by DiOC6.
Magnification: 100x
Back to Gallery