Head HTML-
Hippocampus
Submitted by: Mr Benjamin Barti
Institute of Experimental Medicine  Hungarian Academy of Sciences
In the image we can see manually filled interneurons (orange) in the hippocampus stratum radiatum layer of a mouse brain. These neurons forms so-called baskets with their axon boutons mostly on the somas of the pyramidal cells which nuclei (with all other cell nuclei) were labeled with DAPI (cyane). Image was captured with confocal microscope.
Magnification: 10x
Back to Gallery