Head HTML-
Porcine Carotid Artery
Submitted by: Siobhan Dunphy
Trinity College Dublin
Porcine smooth muscle cells were seeded on a decellularised porcine carotid artery and cultured for 7 days in DMEM with 10% FBS. The collagen fibres of the artery were stained with a CNA35 collagen binding probe (red) and the cells were stained with Phalloidin (green). The image was taken using a Leica SP8 scanning confocal microscope.
Magnification: 20x
Back to Gallery