Blueberry Root
Submitted by: Dr. Paola Beraldo, University of Udine, Department of Food Science
A section of the underground stem runner of blueberry (Vaccinium myrtillus), the tissue shown in the image is the xylem (wood) and pith specifically. It was stained using toluidine blue 0.1%. The picture was shot with a Nikon camera (DS camera control unit-DS-L3) in phase contrast microscopy (Leica DMRB).
Maginification: 100x
Back to main gallery page