Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Protein of the Month
Protein of the Month, February: Tev Protease, Expressed in E. Coli
Origin: Tobacco Etch Virus (TEV) causes dwarfing of plants and wilting of their leaves in plants such as tobacco or tomato.. The virus proteins are organized in a polyprotein that is specifically cleaved.
Action: The protease specificity helps the virus process its polyprotein to functional units. The specificity is also useful to cleave engineered recombinant fusion proteins.
How: The TEV protease is highly specific to recognition sequence Glu-Asn-Leu-Tyr-Phe-Gln/Gly.
Did you know? TEV protease with GST and His tags allows both the protease and the cleaved target tag to be removed at the same time.
  Learn More:
Return to mainpage