Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Protein of the Month
Protein of the Month, June: 𝛽-Casein from Bovine Milk
General Description 𝛽-Casein is part of a family of proteins called phosphoproteins, found in mammalian milk.
Application: Casein has numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzymatic activity from SDS extracted samples and as a substrate for protease and kinase assays.
Did you know? The use of casein was used in prehistoric times in cave-paintings. Since the 1960s, acrylic paints have become the popular choice.
  More Information:
Return to mainpage