Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Protein Of The Month
May: Mucin
Origin: Mucus forms the protective cover for all epithelial surfaces. The major structural component of mucus is gel-forming mucins.
Application: It was used in a study to evaluate sputum smears concentrated by cyto-centrifugation for detection of acid-fast bacilli.
How: Mucins are a diverse family of densely glycosylated proteins. The highly glycosylated properties of mucins make them resistant to proteolysis. This means they are able to hold water, giving them the gel-like properties found in mucosal barriers.
Did you know: The mucus barrier that protects the human respiratory system is so effective it prevents not only bacteria/ foreign materials but also inhaled drug therapeutics from reaching their targets. Researchers have conducted a study with newly designed 'DNA-loaded nanoparticles' that can cross the mucus barrier in the lungs. Visit the John Hopkins website to learn more.
Learn More:
Return to mainpage