Protein of The Month
October: Benzonase Nuclease
Origin: The enzyme was isolated from the bacterium Serratia marcescens and then genetically engineered. This is an example of how useful properties can be extracted even from a source that is typically a pathogen.
Action: Benzonase nuclease can be used to degrade all forms of DNA and RNA while having no proteolytic activity. It also reduces viscosity in protein extracts and prevents cell clumping.
How: Enzymatic activity is often evaluated and tested on salmon sperm DNA.
Did you know? Benzonase is one of the few nucleases that hydrolyzes all forms of DNA and RNA.
  Learn More:
Return to mainpage