-
Promotion only available with the Promotion Code HFI until February 29, 2012
(European Customers only)
The BactiZapper uses infrared heating to produce 815 °C (1500 °F). It completely sterilizes microorganisms in 5-7seconds. The BactiZapper uses well-proven technology to provide instant sterilization of platinum inoculating loops, needles, glass tube/pipette mouths and various metal & borosilicate glass instruments, commonly used in microbiology research.
It is extremely compact, convenient and can be used in anaerobic and aerobic chambers.
| Product No. | Description | Discount [%] | Add to Cart |
| Z690643-1EA | BactiZapper™ infrared microsterilizer, AC input 115 V | 20 | |
| Z690651-1EA | BactiZapper™ infrared microsterilizer, AC input 230 V (EU & UK plug) | 20 | |
| Z690686-1EA | BactiZapper™ slide dryer attachment for 3 slides | 20 | |