|
LC-MS Solvent
Save 60%
 Promotion only available with the Promotion Code 993 until August 31, 2011
Fluka LC-MS solvents are specifically formulated to have a low content of alkaline impurities, such as calcium, magnesium, potassium and sodium that can interfere in the analysis by forming artifacts with the analyte. CHROMASOLV® LC-MS solvents are run through specific UV-spectroscopic quality +B7control tests to guarantee CHROMASOLV® specifications.
| Product No. |
Description |
Add to Cart |
| 34965-1L |
2-Propanol LC-MS Chromasolv™ |
|
| 34965-2.5L |
2-Propanol LC-MS Chromasolv™ |
|
| 34965-4X2.5L |
2-Propanol LC-MS Chromasolv™ |
|
| 34965-6X1L |
2-Propanol LC-MS Chromasolv™ |
|
| 34966-1L |
Methanol, LC-MS CHROMASOLV®, ≥99.9% |
|
| 34966-2.5L |
Methanol, LC-MS CHROMASOLV®, ≥99.9% |
|
| 34966-4X2.5L |
Methanol, LC-MS CHROMASOLV®, ≥99.9% |
|
| 34966-4X4L* |
Methanol, LC-MS CHROMASOLV®, ≥99.9% |
|
| 34966-6X1L |
Methanol, LC-MS CHROMASOLV®, ≥99.9% |
|
| 34967-1L |
Acetonitrile LC-MS CHROMASOLV®, ≥99.9% |
|
| 34967-2.5L |
Acetonitrile LC-MS CHROMASOLV®, ≥99.9% |
|
| 34967-250ML |
Acetonitrile LC-MS CHROMASOLV®, ≥99.9% |
|
| 34967-4X2.5L |
Acetonitrile LC-MS CHROMASOLV®, ≥99.9% |
|
| 34967-4X4L* |
Acetonitrile LC-MS CHROMASOLV®, ≥99.9% |
|
| 34967-6X1L |
Acetonitrile LC-MS CHROMASOLV®, ≥99.9% |
|
| 34972-1L-R |
Ethyl Acetate LC-MS CHROMASOLV®, ≥99.7% |
|
| 34972-2.5L-R |
Ethyl Acetate LC-MS CHROMASOLV®, ≥99.7% |
|
| 34986-1L |
Hexane - LC-MS CHROMASOLV®, ≥97% |
|
| 34986-2.5L |
Hexane - LC-MS CHROMASOLV®, ≥97% |
|
| 34999-1L |
Heptane - LC-MS CHROMASOLV®, ≥99% |
|
| 34999-2.5L |
Heptane - LC-MS CHROMASOLV®, ≥99% |
|
| 39253-1L-R |
WATER, LC-MS CHROMASOLV® |
|
| 39253-4X4L-R* |
WATER, LC-MS CHROMASOLV® |
|
| Back to Top |
* US-only