|
Molecular Biology Media & Agars
Save 60%
Promotion only available with the Promotion Code T95 until December 31, 2010 (European Customers only)
| Product No. |
Description |
Add to Cart
|
| LB Agar |
| L2897-1KG |
powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L |
|
| L2897-250G |
powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L |
|
| LB Agar |
| L7025-100TAB |
tablet, 1.68 g per tablet, dissolve the tablet in 50 mL of deionized water. |
|
| L7025-500TAB |
tablet, 1.68 g per tablet, dissolve the tablet in 50 mL of deionized water. |
|
| LB Broth |
| L3022-1KG |
powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L |
|
| L3022-250G |
powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L |
|
| LB Broth |
| L7275-100TAB |
tablet, 1.1 g per tablet. Dissolve the tablet in 50 mL of water. |
|
| L7275-500TAB |
tablet, 1.1 g per tablet. Dissolve the tablet in 50 mL of water. |
|
| LB Agar EZMix™ Powder |
| L7533-1KG |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L |
|
| L7533-6X500ML |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L |
|
| LB Broth EZMix™ Powder |
| L7658-2x5L |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L |
|
| L7658-6X500ML |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L |
|
| Terrific Broth, modified |
| T0918-1KG |
powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L |
|
| T0918-250G |
powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L |
|
| Terrific Broth, Modified EZMix™ Powder |
| T9179-1KG |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L |
|
| T9179-2X5L |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L |
|
| T9179-6X500ML |
granulated, dust free powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L |
|
| YPD Broth |
| Y1375-1KG |
Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L |
|
| Y1375-250G |
Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L |
|
| YPD Agar |
| Y1500-250G |
Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L, Agar 15 g/L |
|
| Y1500-1KG |
Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L, Agar 15 g/L |
|
| Back to Top |