Sigma-Aldrich
Europe > Special Offers - Europe > Molecular Biology Media and Agars
Molecular Biology Media & Agars
Molecular Biology Media & Agars
Save 60%
Promotion OverviewPromotion only available with the Promotion Code T95 until December 31, 2010
(European Customers only)
Product No. Description
Add to Cart
LB Agar
L2897-1KG powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L
L2897-250G powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L
LB Agar
L7025-100TAB tablet, 1.68 g per tablet, dissolve the tablet in 50 mL of deionized water.
L7025-500TAB tablet, 1.68 g per tablet, dissolve the tablet in 50 mL of deionized water.
LB Broth
L3022-1KG powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L
L3022-250G powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L
LB Broth
L7275-100TAB tablet, 1.1 g per tablet. Dissolve the tablet in 50 mL of water.
L7275-500TAB tablet, 1.1 g per tablet. Dissolve the tablet in 50 mL of water.
LB Agar EZMix™ Powder
L7533-1KG granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L
L7533-6X500ML granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L, Agar 15 g/L
LB Broth EZMix™ Powder
L7658-2x5L granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L
L7658-6X500ML granulated, dust free powder, Tryptone (pancreatic digest of casein) 10 g/L, Yeast extract 5 g/L, NaCl 5 g/L
Terrific Broth, modified
T0918-1KG powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L
T0918-250G powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L
Terrific Broth, Modified EZMix™ Powder
T9179-1KG granulated, dust free powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L
T9179-2X5L granulated, dust free powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L
T9179-6X500ML granulated, dust free powder, Tryptone (pancreatic digest of casein) 12 g/L, Yeast extract 24 g/L, K2HPO4 9.4 g/L, KH2PO4 2.2 g/L
YPD Broth
Y1375-1KG Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L
Y1375-250G Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L
YPD Agar
Y1500-250G Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L, Agar 15 g/L
Y1500-1KG Bacteriological peptone 20 g/L, Yeast extract 10 g/L, Glucose 20 g/L, Agar 15 g/L