-
Please mention Promotion Code SBU when ordering, valid until February 29, 2012
OPTI-GUARD® 1 mm long guard columns pack the power of a guard column into an auto-adjusting, finger-tight design that is no larger than a typical HPLC fitting. They are the perfect choice for effective column protection of any silica-based analytical column without measurable impact on your chromatography. Designed for use with analytical (4.6 mm, 3.0 mm I.D.) and narrow bore (2.1 mm, 1.0 mm I.D.) columns, the patented floating stem design automatically adjusts to any manufactures tube stop depth for a zero-dead volume connection every time. In addition, the OPTI-GUARD 3mm guard system offers greater capacity and more rugged protection. | Product No. | Description | Discount [%] | Add to Cart |
| 51188-U | Opti-Guard 3mm Peek/SS Holder | 20 | |
| 51177-U | PK5 Opti-Guard 1mm Guard Column C18 (Violet Label) | 20 | |
| 51183-U | PK5 Opti-Guard 1mm Guard Column C18, Biocompatible | 20 | |
| 51184-U | PK5 Opti-Guard 1mm Guard Column C8 | 20 | |
| 51185-U | PK5 Opti-Guard 1mm Guard Column Phenyl | 20 | |
| 51178-U | PK5 Opti-Guard 1mm Guard Column Silica (Orange Label) | 20 | |
| 51179-U | PK5 Opti-Guard 1mm Guard Column CN (Blue Label) | 20 | |
| 51180-U | PK5 Opti-Guard 1mm Guard Column Anion Exchange (Black Label) | 20 | |
| 51181-U | PK5 Opti-Guard 1mm Guard Column Cation Exchange (White Label) | 20 | |
| 51187-U | PK5 Opti-Guard 1mm Guard Column Amino NH2 | 20 | |
| 51191-U | PK3 Opti-Guard 3mm Cartridge C18 | 20 | |
| 51193-U | PK3 Opti-Guard 3mm Cartridge C8 | 20 | |
| 51194-U | PK3 Opti-Guard 3mm Cartridge Amino (NH2) | 20 | |
| 51196-U | PK3 Opti-Guard 3mm Cartridge Silica | 20 | |
| Back to Top | |||