-
Please mention Promotion Code HFI when ordering, valid until February 29, 2012
(European Customers only)
| Product No. | Description | Discount [%] | Add to Cart |
| Sigma’s prestained markers contain specific dye-conjugated proteins. Single and multiple color markers are available. ColorBurst™ wide range markers provide a rainbow of brilliantly colored bands in a ready-to-use format. | |||
| C6210-1VL | Color Marker Ultra-low Range (M.W. 1,060-26,600) | 33 | |
| C1992-1VL | ColorBurst™ Electrophoresis Marker, mol wt 8,000-220,000Da | 33 | |
| C3817-1EA | Nalgene® round carboy, with handles, nominal capacity 20L (5.5gal), low-density polyethylene | 33 | |
| SDS7B2-1VL | Prestained Molecular Weight Marker, mol wt 26,600-180,000Da | 33 | |
| Sigma’s unstained markers yield well-defined bands amenable to most protein electrophoresis staining methods. The low molecular weight ranges of M3546 and MWSDS17S are best used in Tris-Tricine SDS-PAGE applications. All other unstained markers are suited for use in Laemmli SDS-PAGE applications. | |||
| MWSDS17S-1VL | Peptide Molecular Weight Marker, mol wt 2,500-17,000Da | 33 | |
| S8320-1VL | SigmaMarker™, High Range, mol wt 36,000-200,000Da | 33 | |
| S8320-10VL | SigmaMarker™, High Range, mol wt 36,000-200,000Da | 33 | |
| M3913-1VL | SigmaMarker™, low range, mol wt 6,500-66,000Da | 33 | |
| M3913-10VL | SigmaMarker™, low range, mol wt 6,500-66,000Da | 33 | |
| S8445-1VL | SigmaMarker™, Wide Range, mol wt 6,500-200,000Da | 33 | |
| S8445-10VL | SigmaMarker™, Wide Range, mol wt 6,500-200,000Da | 33 | |
| M3546-1VL | Ultra-low Range Molecular Weight Marker (M.W. 1,060-26,600) | 33 | |
| Sigma’s specialty markers are a set of molecular weight markers, each with specific attributes making it unique. Special attention should be paid when selecting one of the specialty markers to ensure it will meet the requirements of the intended application. | |||
| B2787-1VL | Biotinylated Molecular Weight Marker, mol wt 6,500-180,000Da | 33 | |
| F3526-1VL | Fluorescent Molecular Weight Marker, mol wt 20,000-200,000Da | 33 | |
| MWND500-1KT | Kit for Molecular Weights 14,000-500,000 Non-denaturing | 33 | |
| M0671-.5ML | Recombinant Molecular Weight Marker, mol wt 15,000-150,000Da | 33 | |
| M5505-1VL | Silver Stain High Range Molecular Weight Marker (M.W. 29,000-116,000) | 33 | |