Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Europe > Special Offers - Europe > SLB™-5ms Capillary Columns
SLB™ 5ms Capillary Columns
Save 30%
Promotion Overview
Please mention Promotion Code SBU when ordering, valid until February 29, 2012
GC Capillary ColumnsToday’s chromatographers require GC-MS columns with consistently low bleed characteristics. High column bleed may interfere with proper mass spectral identification of analytes and may also foul the MS source. For these reasons, capillary columns that exhibit very low levels of bleed are desirable. Supelco SLB™-5ms (Supelco Low Bleed) capillary columns offer:
• Cleaner Mass Spectra
• Very low bleed even at 325 °C
• Shorter analysis times
• Greater column-to-column reproducibility
Product No. Description Discount [%] Add to Cart
28465-U FS CAP SLB™-5ms 10m 0.10mm 0.10µm 30
28466-U FS CAP SLB™-5ms 15m 0.10mm 0.10µm 30
28467-U FS CAP SLB™-5ms 30m 0.25mm 0.10µm 30
28469-U FS CAP SLB™-5ms 15m 0.25mm 0.25µm 30
28471-U FS CAP SLB™-5ms 30m 0.25mm 0.25µm 30
28472-U FS CAP SLB™-5ms 60m 0.25mm 0.25µm 30
28473-U FS CAP SLB™-5ms 30m 0.25mm 0.50µm 30
28474-U FS CAP SLB™-5ms 60m 0.25mm 0.50µm 30
28476-U FS CAP SLB™-5ms 30m 0.25mm 1.0µm 30
28482-U FS CAP SLB™-5ms 30m 0.32mm 0.25µm 30
28484-U FS CAP SLB™-5ms 30m 0.32mm 0.50µm 30
28487-U FS CAP SLB™-5ms 30m 0.32mm 1.0µm 30
28513-U FS CAP SLB™-5ms 30m 0.20mm 0.20µm 30
28532-U FS CAP SLB™-5ms 30m 0.32mm 0.32µm 30
28541-U FS CAP SLB™-5ms 30m 0.53mm 0.50µm 30
28542-U FS CAP SLB™-5ms 15m 0.53mm 0.50µm 30
28557-U FS CAP SLB™-5ms 15m 0.32mm 0.25µm 30
28559-U FS CAP SLB™-5ms 30m 0.53mm 1.0µm 30
28564-U FS CAP SLB™-5ms 20m 0.18mm 0.18µm 30
28566-U FS CAP SLB™-5ms 12m 0.18mm 0.30µm 30
28575-U FS CAP SLB™-5ms 30m 0.18mm 0.30µm 30
28576-U FS CAP SLB™-5ms20m 0.18mm 0.36µm 30
28577-U FS CAP SLB™-5ms 15m 0.25mm 0.50µm 30
28597-U FS CAP SLB™-5ms 15m 0.32mm 0.50µm 30