Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Europe > Special Offers - Europe > Supel Select
Supel™-Select HLB SPE
Save 20%
Promotion Overview
Please mention Promotion Code SBU when ordering, valid until February 29, 2012
Supel-SelectGet 20% Discount for Supel™-Select HLB SPE
Supel™-Select HLB SPE is a hydrophilic modified styrene-based polymer developed for solid phase extraction (HLB: Hydrophilic Lipophilic Balance). Due to its hydrophilic/lipophilic properties, this phase extracts a wide variety of analytes, for example from body fluids. Also available in 96-well plates.
To read more, please see: sigma-aldrich.com/supel-select
Product No. Description Discount [%] Add to Cart
54186-U Supel™-Select HLB SPE Tube 1g/20ml Pk20 20
54183-U Supel™-Select HLB SPE Tube 200mg/6ml Pk30 20
575661-U Supel™-Select HLB SPE 96-Well Plate 30mg/well Pk1 20
575662-U Supel™-Select HLB SPE 96-Well Plate 60mg/well Pk1 20
54181-U Supel™-Select HLB SPE Tube 30mg/1ml Pk100 20
54184-U Supel™-Select HLB SPE Tube 500mg/12ml Pk20 20
54182-U Supel™-Select HLB SPE Tube 60mg/3ml Pk50 20