|
Supel-Tips
Save 20%
 Promotion only available with the Promotion Code 993 until August 31, 2011
Pipette tips are convenient and useful tools for extraction, concentration, and/or purification of complex bio-molecules in very small sample volumes. The 10μL pipette tips incorporate a SPE bed immobilized by a proprietary attachment process preventing any release of particles.
| Product No. |
Description |
Add to Cart |
| 54227-U |
Supel-Tips Carbon Pipette Tips, Pk96 |
|
| 54263-U |
Supel-Tips Ti Pipette Tips, Pk96 |
|
| 54266-U |
Supel-Tips Zr Pipette Tips, Pk96 |
|
| TPSC18-96EA |
Supel-Tips C18 Pipette Tips, Pk96 |