-
Promotion only available with the Promotion Code HFI until February 29, 2012
(European Customers only)
Protran nitrocellulose (NC) membranes are the most frequently specified transfer media in the world for a wide range of applications. Protran membranes are manufactured using 100% pure nitrocellulose to ensure the highest binding capacity possible. Other membranes referred to as ‘nitrocellulose’ may actually contain large amounts of cellulose acetate which will lower the protein binding capacity. Protran membranes have the best handling strength of all pure nitrocellulose membranes. They are compatible with a variety of detection methods, including isotopic, chemiluminescent (luminol-based), colorimetric and fluorescent. Unlike PVDF membranes, Protran nitrocellulose does not require a methanol pre-wetting step. This makes Protran the membrane of choice for proteins which prefer aqueous environments. Prior to transfer, the membrane is simply wetted in water, and then placed in the transfer buffer. No other pre-treatment steps are necessary.
Available as rolls, sheets or circles.
| Product No. | Description | Discount [%] | Add to Cart |
| Z613630-1ea | Whatman® Protran® nitrocellulose membranes Rolls - BA85, pore size 0.45 μm, W × L 30 cm × 3 m | 20 | |
| Z613649-5ea | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 30 cm × 60 cm | 20 | |
| Z613657-1ea | Whatman® Protran® nitrocellulose membranes Rolls - BA83, pore size 0.2 μm, W × L 30 cm × 3 m | 20 | |
| Z613665-25EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 20 cm × 20 cm | 20 | |
| Z613673-50EA | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 82 mm | 20 | |
| Z613681-50EA | Whatman® Protran® nitrocellulose membranes Circles - BA85 (5 mm grid), pore size 0.45 μm, diam. 82 mm | 20 | |
| Z613703-25EA | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 132 mm | 20 | |
| Z670618-50EA | Whatman® Protran® nitrocellulose membranes Circles - BA83, pore size 0.2 μm, diam. 82 mm | 20 | |
| Z670626-50ea | Whatman® Protran® nitrocellulose membranes Circles - BA83, pore size 0.2 μm, diam. 132 mm | 20 | |
| Z670634-100EA | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 25 mm | 20 | |
| Z670642-1000EA | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 25 mm | 20 | |
| Z670650-50ea | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 82.5 mm | 20 | |
| Z670669-50EA | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 87 mm | 20 | |
| Z670677-50EA | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 132 mm | 20 | |
| Z670685-25ea | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 137 mm | 20 | |
| Z670693-50ea | Whatman® Protran® nitrocellulose membranes Circles - BA85, pore size 0.45 μm, diam. 137 mm | 20 | |
| Z670715-10EA | Whatman® Protran® nitrocellulose membranes Sheets - BA79, pore size 0.1 μm, L × W 10.2 cm × 13.3 cm | 20 | |
| Z670731-5ea | Whatman® Protran® nitrocellulose membranes Sheets - BA79, pore size 0.1 μm, L × W 20 cm × 20 cm | 20 | |
| Z670758-25EA | Whatman® Protran® nitrocellulose membranes Sheets - BA79, pore size 0.1 μm, L × W 20 cm × 20 cm | 20 | |
| Z670766-5ea | Whatman® Protran® nitrocellulose membranes Sheets - BA79, pore size 0.1 μm, L × W 33 cm × 56 cm | 20 | |
| Z670774-10EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 10.2 cm × 13.3 cm | 20 | |
| Z670790-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 15 cm × 15 cm | 20 | |
| Z670804-10EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 15 cm × 20 cm | 20 | |
| Z670812-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 20 cm × 20 cm | 20 | |
| Z670820-5ea | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 25 cm × 25 cm | 20 | |
| Z670839-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 30 cm × 60 cm | 20 | |
| Z670847-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA83, pore size 0.2 μm, L × W 33 cm × 56 cm | 20 | |
| Z670855-10EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 10.2 cm × 13.3 cm | 20 | |
| Z670863-10EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 6.3 cm × 22.8 cm | 20 | |
| Z670871-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 15 cm × 15 cm | 20 | |
| Z670898-25EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 15 cm × 15 cm | 20 | |
| Z670901-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 20 cm × 20 cm | 20 | |
| Z670928-25EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 20 cm × 20 cm | 20 | |
| Z670936-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 25 cm × 25 cm | 20 | |
| Z670944-5EA | Whatman® Protran® nitrocellulose membranes Sheets - BA85, pore size 0.45 μm, L × W 33 cm × 56 cm | 20 | |
| Z670952-1EA | Whatman® Protran® nitrocellulose membranes Rolls - BA79, pore size 0.1 μm, W × L 30 cm × 3 m | 20 | |
| Z670960-1EA | Whatman® Protran® nitrocellulose membranes Rolls - BA83, pore size 0.2 μm, W × L 15 cm × 3 m | 20 | |
| Z670979-1EA | Whatman® Protran® nitrocellulose membranes Rolls - BA83, pore size 0.2 μm, W × L 20 cm × 3 m | 20 | |
| Z670987-1EA | Whatman® Protran® nitrocellulose membranes Rolls - BA85, pore size 0.45 μm, W × L 15 cm × 3 m | 20 | |
| Z670995-1EA | Whatman® Protran® nitrocellulose membranes Rolls - BA85, pore size 0.45 μm, W × L 20 cm × 3 m | 20 | |
| Back to Top | |||