Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Environmental Testing – Organics
Sample preparation involves partitioning organic pollutants into headspace through a purge process (for volatile organics) or into an organic solvent through an extraction process (for non-volatile organics). A cleanup step to remove interferences and/or a concentration step to increase method sensitivity is usually performed. Analysis typically uses gas chromatography (GC) or high performance liquid chromatography (HPLC).
  • Sample Collection
  • Sample Preparation
  • Sample Analysis