サンプル前処理&精製
新規脂質除去剤 Z-Sep
仕様
修飾の異なる2種類の充填剤(Z-SepとZ-Sep+があります。)
Z-Sep
シリカゲル(Si)にジルコニア(ZrO)を化学結合
Z-Sep+
シリカゲル(Si)にジルコニア(ZrO)とC18を化学結合
ジルコニア(左)とC18(右)の保持メカニズム
水酸基はZ-Sepのジルコニア基とのルイス酸-塩基相互作用により保持
C18との疎水性相互作用により保持
Z-Sepは、C18と組み合わせることにより、あるいは、Z-Sep+を単独で用いることにより、全てのオレインを除去可能。同様にモノ-、ジ-、トリグリセリドも除去可能。
Z-Sepの色素除去能
Z-Sepには、カーボンにはない色素除去能が認められ、特にPSAにはない色素(緑色、黄色)除去能もある事が示唆されました。
製品リスト
品名  入数 CAT.NO.
dSPE (dispersive SPE) QuEChERS法用
Z-Sep dSPE CLEAN-UP TUBE (2 mL遠沈管)
Z-Sep:75 mg
100 55411-U
Z-Sep+ dSPE CLEAN-UP TUBE (2 mL遠沈管)
Z-Sep+:75 mg
100 55408-U
Z-Sep dSPE CLEAN-UP TUBE (12 mL遠沈管)
Z-Sep:500 mg
50 55403-U
Z-Sep+ dSPE CLEAN-UP TUBE (12 mL遠沈管)
Z-Sep+:500 mg
50 55296-U
Z-Sep/C18 dSPE CLEAN-UP TUBE (2 mL遠沈管)
Z-Sep:20 mg、C18:50 mg
100 55284-U
Z-Sep/C18 dSPE CLEAN-UP TUBE (12 mL遠沈管)
Z-Sep:120 mg、C18:300 mg
50 55401-U
Z-Sep/MgSO4 dSPE CLEAN-UP TUBE (2 mL遠沈管)
Z-Sep:50 mg、MgSO4:150 mg
100 55417-U
Z-Sep/MgSO4 dSPE CLEAN-UP TUBE (12 mL遠沈管)
Z-Sep:300 mg、MgSO4:900 mg
50 55407-U
Z-Sep+/MgSO4 dSPE CLEAN-UP TUBE (2 mL遠沈管)
Z-Sep+:50 mg、MgSO4:150 mg
100 55414-U
Z-Sep+/MgSO4 dSPE CLEAN-UP TUBE (12 mL遠沈管)
Z-Sep+:300 mg、MgSO4:900 mg
50 55406-U
シリンジ型
Z-Sep 30 mg/1 mL   100 55261-U
200 55276-U
Z-Sep 500 mg/6 mL 30 55267-U
Z-Sep+ 500mg/6mL   
30 52972-U
Z-Sep(55418-U)120mgとC18(52600-U)300mgのブレンド 30 54282-U
充填剤
Z-Sep (20 μm, 120 Å, 350 m²/g、ZrO2%=3.5) 20 g 55418-U
Z-Sep+ (50 μm, 70 Å, 480 m²/g、ZrO2%=4.5/C18%=16) 20 g 55299-U
関連製品
50 mL 遠心分離用エンプティチューブ(抽出操作用) 50 55248-U
Supel is a trademark of Sigma-Aldrich Co. LLC
申込みフォーム
シリンジ型、QuEChERSの無料サンプル配布中
PDF
Z-Sepを使用した分析アプリケーション集です
申込みフォーム
分析でお困りの方
お気軽にご相談ください
参考文献
  • Tomasz K. et al."Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry -honeybee poisoning incidents"Journal of Chromatography A, 1435 (2016) 100–114
  • Darina L. et al."A novel strategy for the determination of polycyclic aromatic hydrocarbon monohydroxylated metabolites in urine using ultra-high-performance liquid chromatography with tandem mass spectrometry"Analytical and Bioanalytical Chemistry(2016)
  • José F. H. et al."Method optimization and validation for the determination of eight sulfonamides in chicken muscle and eggs by modified QuEChERS and liquid chromatography with fluorescence detection"Journal of Pharmaceutical and Biomedical Analysis 124 (2016) 261–266
  • Bruce D. M. et al. "Development of an Automated Column Solid-Phase Extraction Cleanup of QuEChERS Extracts, Using a Zirconia-Based Sorbent, for Pesticide Residue Analyses by LC-MS/MS"J. Agric. Food Chem. (2015) 63, 5107−5119
  • 吉岡直樹、林 幸子、稲田忠明 "かんきつ類における7種防かび剤の迅速分析法の検討" 食衛誌 Vol. 56, No. 5(2015)
  • P. Parrilla Vázquez et al."A sensitive and efficient method for routine pesticide multiresidue analysis in bee pollen samples using gas and liquid chromatographycoupled to tandem mass spectrometry"Journal of Chromatography A, 1426 (2015) 161–173
  • Stanisław W. et al.”Two-step dispersive-solid phase extraction strategy for pesticide multiresidue analysis in a chlorophyll-containing matrix by gaschromatography–tandem mass spectrometry”Journal of Chromatography A, 1412 (2015) 22–32
  • Pol H. et al. "A quick, easy, cheap, effective, rugged and safe extraction method followed by liquid chromatography-(Orbitrap) high resolution massspectrometry to determine benzotriazole, benzothiazole andbenzenesulfonamide derivates in sewage sludge"Journal of Chromatography A, 1339 (2014) 34–41
  • Marta P. et al. "Determination of illegal dyes in eggs by liquid chromatography -tandem mass spectrometry"Bull Vet Inst Pulawy 58(2014) 247-253
  • Yelena S.et al."Multi-class, multi-residue analysis of pesticides, polychlorinated biphenyls, polycyclic aromatic hydrocarbons, polybrominated diphenyl ethers and novel flame retardants in fish using fast, low-pressure gas chromatography–tandem mass spectrometry"Analytica Chimica Acta 758 (2013) 80– 92
  • Łukasz R. et al. "Determination of pesticide residues in high oil vegetal commodities by using various multi-residue methods and clean-ups followed byliquid chromatography tandem mass spectrometry"Journal of Chromatography A, 1304 (2013) 109– 120
  • Nariyasu M. et al. "Identification and elimination of ion suppression in the quantitative analysis of sirolimus in human blood by LC/ESI-MS/MS"Journal of Chromatography B, 879 (2011) 968–974
申込みフォーム
シリンジ型、QuEChERSの無料サンプル配布中
PDF
Z-Sepを使用した分析アプリケーション集です
申込みフォーム
分析でお困りの方
お気軽にご相談ください