PCR産物の生成 in vitro転写反応のテンプレートはRNAポリメラーゼプロモーター配列を含んだ、ターゲットに特異的なプライマーか、ベクターバックボーンに特異的なプライマーを用いたクローンからのcDNAのPCR増幅により生成する。PCR産物は配列検証済みで、増幅領域はDeqor siRNAデザインプログラムに基づいた高効率のsiRNAターゲット配列を含みます。MISSION esiRNAはターゲット遺伝子の様々な転写をカバーするようデザインされています。
In vitro 転写反応とロングdsRNAの作成 cDNAからin vitro 転写反応によってPCR産物を作成します。RNAポリメラーゼでdsRNAを作成し、2つの1本鎖RNAストランドを酵素消化前にアニールします。
Slabicki M., et al. (2010) A Genome-Scale DNA Repair RNAi Screen Identifies SPG48 as a Novel Gene Associated with Hereditary Spastic Paraplegia. PLoS Biol 8(6): e1000408. doi:10.1371/journal.pbio.1000408. Paper (2.22 Mb PDF)
Collinet, et al. Systems survey of endocytosis by multiparametric image analysis. Nature 464, 243-249 (2010). Abstract
Theis, M. et al. Comparative profiling identifies C13orf3 as a component of the Ska complex required for mammalian cell division. EMBO J. 28, 1453-65 (2009). Abstract
Ding, L. et al. A Genome-Scale RNAi Screen for Oct4 Modulators Defines a Role of the Paf1 Complex for Embryonic Stem Cell Identity. Cell Stem Cell. 9, 403-15 (2009). Abstract
Kittler, R. et al. Genome-scale RNAi profiling of cell division in human tissue culture cells. Nat. Cell Biol. 9, 1401-12 (2007). Abstract
Kittler, R. et al. An endoribonuclease-prepared siRNA screen in human cells identifies genes essential for cell division. Nature 432, 1036-40 (2004). Abstract
Additional esiRNA Literature References
Stewart, G. S. et al. The RIDDLE syndrome protein mediates a ubiquitin-dependent signaling cascade at sites of DNA damage. Cell 136, 420-34 (2009). Abstract
Lawo, S. et al. HAUS, the 8-Subunit Human Augmin Complex, Regulates Centrosome and Spindle Integrity. Curr Biol., 19, 816-26 (2009). Abstract
Fazzio, T. G. et al. An RNAi screen of chromatin proteins identifies Tip60-p400 as a regulator of embryonic stem cell identity. Cell 2008 134, 162-74 (2008). Abstract
Konstantinova, I. et al. EphA-Ephrin-A-mediated beta cell communication regulates insulin secretion from pancreatic islets. Cell 129, 359-70 (2007). Abstract
Nikolova, G. et al. The vascular basement membrane: a niche for insulin gene expression and Beta cell proliferation. Dev Cell. 10, 397-405 (2006). Abstract
Kittler, R. et. al. RNA interference rescue by bacterial artificial chromosome transgenesis in mammalian tissue culture cells. Proc. Natl. Acad. Sci. U.S.A. 102, 2396-401 (2005). Abstract
Liu, W. Y. et al. Efficient RNA interference in zebrafish embryos using synthesized with SP6 RNA polymerase. Dev. Growth Differ. 47(5), 323-31 (2005). Abstract
Yang, D. et al. Short RNA duplexes produced by hydrolysis with Escherichia coli RNase III mediate effective RNA interference in mammalian cells. Proc. Natl. Acad. Sci. U.S.A. 99, 9942-7 (2002). Abstract