Japan > ライフサイエンス > 分子生物学 > PCR関連 > 定量PCR(qPCR) > qPCR テクニカルガイド
定量PCR(qPCR)
qPCR テクニカルガイド
SigmaのqPCR製品におけるコンスタントな実験プロトコール、アプリケーション、キットについてご紹介しています。米国でのサービス内容のため、日本のサービス内容と異なる場合があります。
qPCR technical Guide Cover image
(3.28 Mb PDF)
Table of Contents(セパレートPDF)
  1. 定量PCRとは? (PDF 60kb)
  2. qPCR 検出メソッド (PDF 282kb)
  3. プライマー・プローブデザイン (PDF 404kb)
  4. 機器との適合表 (PDF 198kb)
  5. qPCR アプリケーションガイド (PDF 119kb)
  6. qPCR 最適化 (PDF 863kb)
  7. 試薬選択ガイド (PDF 41kb)
  8. トラブルシューティング (PDF 322kb)
  9. 補足1:Dye毎の蛍光波長特性表 (PDF 41kb)
  10. 補足2:RT-qPCR の最適化 (PDF 357kb)
  11. References (PDF 147kb)
Introduction
The routine study of DNA became practical with the invention of the polymerase chain reaction (PCR) by Kary Mullis in 1983. With the advent of PCR, it was possible to multiply a given DNA segment from complex genetic material millions of times in a few hours using simple equipment. PCR provided researchers with the ability to generate enough genetic material to study gene function and the effects of mutations, offering new possibilities in basic research and diagnostics.
Despite these advances, quantitation of DNA or RNA in cells remained a difficult task until 1993 when Russell Higuchi, et al.,1 introduced real-time, or kinetic, monitoring of DNA amplification. Higuchi’s experiments revealed that the relationship between the amount of target DNA and the amount of PCR product generated after a specific number of amplification cycles is linear. This observation formed the basis for real-time quantitative PCR (qPCR).
Numerous qPCR detection chemistries and instruments are now available to answer a wide range of questions. For instance, qPCR can be used to measure viral load or bacterial pathogens in a clinical sample, to verify microarray data, for allelic discrimination or to determine RNA (via cDNA) copy numbers to analyze the level of gene product in a tissue sample.
The Quantitative PCR Technical Guide from Sigma-Aldrich is intended to provide new users with an introduction to qPCR, an understanding of available chemistries, and the ability to apply qPCR to answer research questions. The guide also contains numerous tips and tools for the experienced qPCR user.
シグマ アルドリッチ ジャパン株式会社
ライフサイエンス事業部
〒140-0002 東京都品川区東品川2-2-24 天王洲セントラルタワー4F
■ 製品に関するお問合せは、弊社テクニカルサポートへ
TEL:03-5796-7330 / FAX:03-5796-7335 / E-mail:sialjpts@sial.com
■ 在庫紹介・ご注文方法に関するお問合せは、弊社カスタマーサービスへ
TEL:03-5796-7320 / FAX:03-5796-7325