| 製品番号 |
製品名 |
備考 |
| カソード材料 |
| 729183 |
Lanthanum nickelate, strontium doped nanopowder |
La1.6Sr0.4NiO4 |
| 729175 |
Lanthanum nickelate nanopowder |
La2NiO4 |
| 704288 |
Lanthanum strontium cobalt ferrite, LSCF 6428 |
La0.6Sr0.4Co0.2Fe0.8O3 |
734632 NEW! |
Lanthanum strontium cobalt ferrite |
La0.6Sr0.4Co0.2Fe0.8O3 |
734640 NEW! |
Lanthanum strontium cobalt ferrite, powder, <0.5 μm particle size |
La0.6Sr0.4FeO3 |
| 704296 |
Lanthanum strontium manganite, LSM-20 |
(La0.80Sr0.20)MnO3 |
| 704261 |
Lanthanum strontium manganite, LSM-35 |
(La0.65Sr0.35)MnO3 |
| 704253 |
Lanthanum strontium cobalt ferrite composite cathode powder, LSCF/GDC |
(Ce0.9Gd0.1)O1.95 : (La0.60Sr0.40)(Co0.20Fe0.80)O3 = 1 : 1 |
| 704245 |
Lanthanum strontium manganite composite cathode powder, LSM-20/YSZ |
(Y2O3)0.08(ZrO2)0.92 : (La0.80Sr0.20)MnO3 = 1 : 1 |
| 704237 |
Lanthanum strontium manganite composite cathode powder, LSM20-GDC10 |
(Ce0.9Gd0.1)O1.95 : (La0.80Sr0.20)MnO3 = 1 : 1 |
734659 NEW! |
Manganese cobalt oxide, spinel, powder, <0.5 μm particle size |
Mn1.5Co1.5O4 |
| アノード材料 |
| 704210 |
Nickel oxide - Cerium samarium oxide for coatings, NiO/SDC, 99+ % trace metals basis |
NiO : (CeO2 - Sm2O3) = 60 : 40 |
| 704229 |
Nickel oxide - Yttria-stabilized zirconia (YSZ) for general applications, NiO/YSZ, 99+ % trace metals basis |
NiO : (Y2O3 – ZrO2) = 60 : 40 |
| 704202 |
Nickel oxide - Yttria-stabilized zirconia (YSZ) for coatings, NiO/YSZ, 99+ % trace metals basis |
NiO : (Y2O3 - ZrO2) = 66 : 34 |
| 203882 |
Nickel(II) oxide, 99.99% trace metals basis |
NiO |
| 電解質 |
| 729191 |
Cerium oxide, praseodymium doped nanopowder |
Ce0.8Pr0.2O2 |
| 572330 |
Cerium(IV) oxide-gadolinium doped (GDC) nanopowder |
Gd 10 mol % as dopant |
734519 NEW! |
Cerium(IV) oxide-gadolinium doped, powder <0.5 μm particle size |
Gd 10 mol % as dopant |
| 572357 |
Cerium(IV) oxide-gadolinium doped (GDC) nanopowder |
Gd 20 mol % as dopant |
734667 NEW! |
Cerium(IV) oxide-gadolinium doped, pellets (for sputtering, diam.×L: 2-3 mm×3-5 mm) |
Gd 20 mol % as dopant |
| 572365 |
Cerium(IV) oxide-samaria doped nanopowder |
SmO2 15 mol % as dopant |
734675 NEW! |
Cerium(IV) oxide-samarium doped, pellets (for sputtering, diam.×L: 2-3 mm×3-5 mm) |
Sm 15 mol % as dopant |
734624 NEW! |
Cerium(IV) oxide-samarium doped, powder , <0.5 μm particle size |
Sm 20 mol % as dopant |
| 572322 |
Zirconium(IV) oxide-yttria stabilized (YSZ) nanopowder |
Y2O3 3 % as stabilizer |
| 572349 |
Zirconium(IV) oxide-yttria stabilized (YSZ) nanopowder |
Y2O3 8 % as stabilizer |
| 464228 |
Zirconium(IV) oxide-yttria stabilized (YSZ) submicron powder |
Y2O3 8 % as stabilizer, 99.9% trace metals basis (purity excludes ~2% HfO2) |
| 634174 |
Cerium(IV)-zirconium(IV) oxide nanopowder, <50 nm particle size (BET), 99.0% trace metals basis |
Ce : Zr = 1: 1 (ZrO2 and CeO2 or an intermediate compound) |
734683 NEW! |
Zirconium(IV) oxide-yttria stabilized, pellets (for sputtering, diam.×L: 2-3 mm×3-5 mm) |
Y2O3 8 % as stabilizer |