Liquid Handling | Microplate Selection
Corning cell culture products offer some of the most advanced surface treatments and culture formats on the market. As a Premier Supply Partner of Corning Life Science products, we offer a complete line of Corning Cell Culture products including: cell culture flasks, cell culture dishes, pipettes, spinner flaks, centrifuge tubes, microplates, roller bottles, cryogenic vials and culture tubes.
Corning Cell Culture Product Categories:
 Corning Low Profile Flask Compared with T175 Flask
|
|
Cell Culture Flasks
- Cell growth areas from: 25 cm² to 1740 cm²
- Innovative surface treatments: CellBIND, tissue-culture, ultra low attachment, non-treated,
- Specialty flasks: low profile, HYPERFlask™, RoboFlask
|
 Corning Petri Dish with Grid
|
|
Cell Culture Dishes
- Manufactured from optically-clear virgin polystyrene
- Innovative surface treatments: Ultra-Web, CellBIND, ultra-low attachment, tissue culture treated
|
 Stripette Serological Pipettes
|
|
Pipettes
- Aspiration, Bacteriological, Pasteur, Serological, and Volumetric pipettes
- Corning Stripette® brand Serological Pipettes
- Anti-drip tips
- Individual wrapped, bulk packaged, or new clean room pack style
- Sterile
|
 Disposable Spinner Flask 500 mL
|
|
Spinner Flasks
- Plastic disposable spinner flasks from 125 mL to 3 L capacity
- Reusable Pyrex spinner flasks from 125 mL to 36 L capacity
- Available with baffles, extra-wide mouth and vertical or angled side-arms
|
 CellSTACK Culture Chamber 10-Stack
|
|
CellSTACK® Culture Chambers
- Cell growth areas from 636 cm² (1 stack) to 25440 cm² (40 Stacks)
- Surface treatments include: CellBIND, tissue culture treated, and ultra-low attachment surface
|
 Centrifuge Tubes
|
|
Centrifuge Tubes
- Centrifuge tubes in Pyrex glass, polyethylene terephthalate (PET) and polypropylene
- Plug seal, screw cap or Centristar cap styles
|
 Spatulas and Microspatulas
|
|
Spatulas and Microspatulas
- Designed for contamination-free samples
- Sterile, RNase/DNase free, non-pyrogenic and antistatic
|
 1536 Well Microplates
|
|
Multiple Well Plates
- 6 to 1536 well microplates
- Surface treatments: Ultra-Web, non-binding, low-binding, non-treated, tissue culture treated, CellBIND, poly-D-lysine
- Available plate styles: low volume, UV, Filtex, PCR, Protein Crystallization, and storage
|
 1700 cm² Expanded Surface Roller Bottle
|
|
1700 cm² Expanded Surface Roller Bottle
- Cell growth areas range from 800 cm² to 1750 cm²
- Cap Styles: Easy Grip, Easy Grip Vented
- Surface treatments: CellBIND, tissue culture treated
- Printed lot numbers,
- Sterile and non-pyrogenic
|
 1000 mL Erlenmeyer Flask
|
|
Erlenmeyer Flasks
- Ideal for shaker culture applications, liquid handling or storage
- Bottle capacities: 125 mL to 3000 mL
- Available with or without baffles
- 26 mm to 46 mm neck diameter
- Cap styles: plug seal or vented caps
- Sterile
|
 Cryogenic Vials
|
|
Cryogenic Vials
- Vial capacity: 1.2 mL to 5.0 mL
- External thread, internal thread, or plug seal caps
- Packaged in boxes, racks or trays
|
 Culture Tubes |
|
Culture Tubes
- Cell culture treated tubes supplied racked
- Sterilized by gamma radiation
- Threaded plug seal caps prevent leakage