Sigma-Aldrich
Labware > Corning Products > Corning Plasticware > Microplate Selection
Labware
Microplate Selection
Cell Culture | Liquid Handling
  • Premium quality Corning® and Costar® brands
  • Expanded product offering
  • Modified catalog numbers
  • Convenient cross-reference list
  • Shopping cart feature for internet ordering
Corning_Logo_For_Web
96 Well Microplates
  • Solid black, white, and clear polystyrene plates
  • Black/white with clear bottom, polystyrene plates
  • Polypropylene plates
  • Polypropylene blocks
384 Well Microplates
  • Solid black, white, and clear polystyrene plates
  • Black/white with clear bottom, polystyrene plates
  • Polypropylene plates/blocks
1536 Well Microplates
24 Well HTS Transwell Microplates
96 Well HTS Transwell Microplates
96 Well HTS Receiver Plate
96 Well HTS Reservoir Plate
96 Well Microplates
Solid black, white, and clear polystyrene plates
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well flat bottom CLS3650, CLS3916, CLS3915,
CLS3361, CLS3590, CLS9018,
CLS3591, CLS9017, CLS3641,
CLS3628, CLS3370, CLS2507,
CLS2509, CLS2503, CLS3665,
CLS3600, CLS3362, CLS3917,
CLS3912, CLS3925, CLS3922,
CLS3596, CLS3977, CLS3598,
CLS3599, CLS3585, CLS3595,
CLS3300, CLS3474
360 10.67 6.86/6.35 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 12.97 3.53 1.27 0.3165 2.26
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well round bottom CLS3366, CLS3797, CLS3360,
CLS3367, CLS3788, CLS3795,
CLS3798, CLS3605, CLS3789,
CLS3792, CLS3799
360 11.3 6.86/6.35 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 13.04 3.53 1.14 N/A 1.76
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well v-bottom CLS3896, CLS3897, CLS3898,
CLS3894, CLS3342, CLS3347
320 11.12 6.86/6.37 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 13.04 4.29* N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well Easy-Wash™ bottom CLS3368, CLS3369 360 10.67 6.86/4.57 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 13.02 3.55 1.27 0.1639 2.26
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 half area flat bottom CLS3686, CLS3688, CLS3690,
CLS3693, CLS3694, CLS3695,
CLS3697, CLS3696, CLS3875
190 10.54 5.00/4.50 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 12.96 2.28 1.27 0.1586 2.39
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well A/2 UV plate CLS3679 205 11.47 5.0/4.5 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 12.49 2.75 0.381 0.1586 2.349
Black/white with clear bottom, polystyrene plates
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well flat bottom CLS3603, CLS3604, CLS3610,
CLS3631, CLS3632, CLS3651,
CLS3666, CLS3667, CLS3903,
CLS3904, CLS3601, CLS3635,
CLS3340
360 10.67 6.86/6.35 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 13.12 3.57 0.5 0.3165 3.03
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well area well flat bottom CLS3880, CLS3881, CLS3882,
CLS3883, CLS3884, CLS3885,
CLS3886, CLS3887, CLS3682,
CLS3679
205 11.47 5.0/4.5 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 12.49 2.75 0.381 0.1586 2.349
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well special optics plate CLS3614, CLS3615 360 10.67 6.86/6.35 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 13.12 2.75 0.127 0.3165 3.403
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well black clear bottom Ultra-Web™ Synthetic Surface CLS3872XX1, CLS3873XX1 366 10.69 6.86/6.35 127.8 85.5 14.2 11.2 14.4
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.1 13.08 3.46 0.295 0.317 3.27
Polypropylene plates
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well flat bottom CLS3364 360 10.67 6.86 (top) 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 130.2 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well round bottom CLS3355, CLS3356, CLS3359,
CLS3365
360 11.3 6.86 (top) 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 12.18 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well v-bottom CLS3357, CLS3363 320 11.13 6.86 (top) 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 13 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well ClearPro round bottom plate CLS3371 360 11.3 6.86/3.22 127.8 85.5 14.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 12.18 N/A N/A N/A –
Polypropylene blocks
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well 0.5 mL v-bottom block CLS3956, CLS3957 500 25.3 6.86(top) 127.8 85.5 27.56 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 24.49 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well 1 mL round bottom block CLS3958, CLS3959 1000 40.28 6.86(top) 127.8 85.1 42.2 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 38.81 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
96 well 2 mL v-bottom block (square well) CLS3960, CLS3961 2000 42.03 8.15 (top width) 127.8 85.9 43.8 11.2 14.3
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
9 6.096 41.35 N/A N/A N/A –
back to top
384 Well Microplates
Solid black, white, and clear polystyrene plates
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well flat bottom CLS3640, CLS3652, CLS3654,
CLS3662, CLS3680, CLS3700,
CLS3701, CLS3702, CLS3703,
CLS3704, CLS3705, CLS3708,
CLS3709, CLS3710, CLS3723
112 11.43 3.63/2.67 (width) 127.8 85.5 14.2 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 6.096 13.15 2.794 1.27 0.0619 1.5
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 low volume round bottom CLS3673, CLS3674, CLS3676,
CLS3677, CLS3678
35 6.58 3.30/2.03 127.8 85.5 14.2 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 6.096 12.84 8.15 0.91 N/A 6.71
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 low flange flat bottom CLS3570, CLS3571, CLS3572,
CLS3573, CLS3574, CLS3575
112 11.43 3.63/2.82 (width) 127.8 85.5 14.2 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 2.41 12.95 2.794 1.4 – 1.37
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 low volume flat bottom CLS3820, CLS3821, CLS3822,
CLS3824, CLS3825, CLS3826
50 9.39 3.30/2.00 127.8 85.5 12.19 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 2.41 10.49 2.79 0.635 0.0314 2.165
Black/white with clear bottom, polystyrene plates
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well flat bottom CLS3653, CLS3655, CLS3663,
CLS3664, CLS3706, CLS3707,
CLS3711, CLS3712, CLS3683
112 11.43 3.63/2.67 (width) 127.8 85.5 14.2 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 6.096 13.15 2.79 0.635 0.0619 2.135
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well clear bottom low volume CLS3540, CLS3542 50 9.39 3.30/2.00 127.8 85.5 12.19 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 2.41 10.49 2.79 0.635 0.0314 2.165
Polypropylene plates/blocks
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well PP round bottom plate CLS3656, CLS3657, CLS3658 95 11.57 3.63 (top width) 127.8 85.5 14.2 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 6.096 12.9 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well PP low volume round bottom CLS3672 35 6.3 3.30/1.98 127.8 85.5 10.0 8.99 12.13
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 2.1 9.24 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well PP round bottom block CLS3964, CLS3965 180 25.11 3.63 (top width) 127.8 85.5 27.8 8.99 12.12
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 6.096 26.71 N/A N/A N/A –
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
384 well PP v-bottom block CLS3342, CLS3347 180 22.4 3.58/3.23 127.3 85.3 24.94 8.94 11.94
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
4.5 2.51 23.42 2.54 1.02 0.322 –
back to top
1536 Well Microplates
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
1536 well solid, round bottom CLS3936, CLS3937 10 5.01 1.69/0.74 127.8 85.5 10.4 7.86 11.0
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
2.25 2.16 8.7 5.41 0.9 N/A 4.49
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
1536 well black, clear bottom CLS3891, CLS3893 12.8 4.8 1.8/1.63 127.8 85.5 10.4 7.86 11.0
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
2.25 2.16 8.9 5.6 0.076 0.0209 5.524
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
1536 well 2 µL "wafer" CLS3851, CLS3852, CLS3853,
CLS3854, CLS3855, CLS3857,
CLS3858
2 1.5 1.19/1.50 127.8 85.5 3.81 7.86 11.01
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
2.25 3.17 3.05 2.31 1.04 0.01884 1.27
  Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
1536 well solid flat bottom CLS3724, CLS3725, CLS3726,
CLS3727, CLS3728, CLS3729
12.8 4.8 1.8/1.63 127.8 85.5 10.4 7.86 11.0
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
2.25 2.16 8.9 5.6 0.076 0.0209 5.524
back to top
24 Well HTS Transwell Microplates
Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
CLS3378, CLS3379, CLS3395,
CLS3396, CLS3397, CLS3398,
CLS3399
1900 – – 127.89 85.6 27.18 – –
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
Distance from Lid to Bottom of Plate
– – – – – – – –
96 Well HTS Transwell Microplates
Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
CLS3374, CLS3380, CLS3381,
CLS3384, CLS3385, CLS3386,
CLS3387, CLS3388, CLS3391,
CLS3392
360 11.811 4.893/4.267 123.748 81.458 13.335 9.23 12.438
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
Distance from Lid to Bottom of Plate
9 7.874 – 4.191 0.01 117.46 13.335 –
96 Well HTS Receiver Plate
Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
CLS3382, CLS3583 360 11.328 10.168 127.762 85.471 14.224 12.004 13.614
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
Distance from Lid to Bottom of Plate
8.999 6.096 14.224 – 1.016 – 14.224 16.193
96 Well HTS Reservoir Plate
Representative Microplate Well
Volume
(µL)
Well Depth (mm) Well Diameter (top/bottom) (mm) Plate Length (mm) Plate Width (mm) Plate Height (mm) A1 Row Offset (mm) A1 Column Offset (mm)
CLS3383 N/A N/A N/A 127.762 85.471 14.224 – –
Well Center-to-center Spacing (mm) Flange or Skirt Height (mm) Stack Height Well Bottom Elevation (mm) Well Bottom Thickness (mm) Well Bottom Area (cm2) Distance to
Bottom of Plate
Distance from Lid to Bottom of Plate
8.999 6.096 14.224 – 1.143 – 14.224 16.193