Glassware Catalog
Tubing Adapters - Sigma-Aldrich Glassware Catalog
Connecting  •  Reducing/Enlarging  •  Septum-Inlet  •  Specialty  •  Tubing  •  Vacuum
On this page: Torion connectionW/ StopcockAccessories
Tubing adapters with Torion connection
  1. Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE- coated sealing ring of the adapter.
  2. Turn the bushing to make an airtight compression seal with the tubing.
  3. Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use.
Tubing
o.d. (in.)
Inner
S.T. Joint
Straight
Catalog Number
With hose barb
Catalog Number
3/16
14/20
3/16
19/22
5/16
14/20
5/16
19/22
5/16
24/40
5/16
29/32
1/2
24/40
1/2
29/32
1/2
34/45
1/2
45/50
Back to Top
Tubing adapters with stopcock
Glass and PTFE stopcocks are threaded for additional safety and have a 2mm bore for 3/16 in. tubing connection and 4mm bore for 5/16 in. and 1/2 in. tubing connections.
Tubing
o.d. (in.)
Inner
S.T. Joint
PTFE stopcock
Catalog Number
Glass stopcock
Catalog Number
3/16
14/20
3/16
19/22
5/16
14/20
5/16
19/22
5/16
24/40
5/16
29/32
1/2
24/40
1/2
29/32
1/2
34/45
1/2
45/50
Back to Top
ACCESSORIES
Solid glass plugs
Replace transfer tubing with this plug to make a leakproof Torion seal.
Plug o.d. (in.)
Catalog Number
3/16
5/16
1/2
Replacement PTFE-coated sealed rings
I.d. (in.)
Catalog Number
3/16
5/16
1/2
Back to Top