Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Labware > Glassware Center > Air-Sensitive Glassware > Inert Gases & Equipment
Air-Sensitive Glassware
Inert Gases & Equipment
Schlenk-Type  •  Storage  •  Septa  •  Inert Gases & Equipment
On this page:
Aldrich inert gasesAldrich RegulatorsMatheson Model 3180 Regulator
Aldrich inert gases
Gases are supplied in convenient, disposable lecture-bottles with a CGA 110F/180M outlet valve.
Order gas regulator below.
Gas
Vol. (L)
Catalog Number
Argon, 99.998+%
57
Nitrogen, 99.998+%
57
Back to Top
Aldrich inert gas regulators for lecture-bottles
al_Z14670P_296 CGA 180 inlet, NPTM outlet connection.
Delivery pressure (psig)
Gauge (psig)
Catalog Number
0 - 100
0 - 150
0 - 15
0 - 30
Back to Top
Matheson Model 3180 inert gas regulators
al_Z40582P For use with standard-size inert gas cylinders.
Specifications:
  • Single-stage
  • Forged brass body/chrome-plated brass bonnet
  • Nylon reinforced neoprene diaphragm
  • Polyurethane seat/nylon seals
  • 3,000 psig max. inlet pressure
  • Temp. range: 0-140°F
  • Outlet needle valve with 1/4in. NPTM connection
CGA connection
Pressure (psig)
Catalog Number
580 (N2, He, Ar)
2 - 15
5 - 125
10 - 200
Back to Top